Skip to Content
Merck
All Photos(1)

Documents

32458

Supelco

Glycarbylamide

PESTANAL®, analytical standard

Synonym(s):

4,5-Imidazoledicarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N4O2
CAS Number:
Molecular Weight:
154.13
Beilstein:
147831
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

NC(=O)c1nc[nH]c1C(N)=O

InChI

1S/C5H6N4O2/c6-4(10)2-3(5(7)11)9-1-8-2/h1H,(H2,6,10)(H2,7,11)(H,8,9)

InChI key

NNWAARLSYSBVPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glycarbylamide may be used as an analytical reference standard for the determination of the analyte in chicken liver extracts and tissues by high performance liquid chromatography (HPLC) with post-column reaction.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Yamamoto et al.
Journal of AOAC International, 82(2), 248-250 (1999-04-07)
A high-performance liquid chromatographic (HPLC) method for determining glycarbylamide (GB) in chicken tissue was developed. GB was extracted with acetonitrile, followed by solid-phase extraction cleanup using a Bond Elut cartridge column with neutral alumina. After the extract had been evaporated
H D Chapman
Veterinary parasitology, 15(1), 11-27 (1984-07-01)
Drug resistance is now recognised as a major cause of the failure of drugs to control coccidiosis in the fowl. In this article, biological, biochemical and genetic aspects of resistance in Eimeria are reviewed and some of the problems that
Yasuharu Mizuno et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 47(6), 293-295 (2007-01-19)
A newly developed HPLC method for determination of glycarbylamide (GB) in chicken liver was applied to other tissues (muscle, fat and kidney). The recoveries of GB in muscle, fat and kidney were 87.2% (CV 0.5), 91.3% (CV 4.7), 79.7% (CV
Noriko Tajima et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 47(6), 254-257 (2007-01-19)
A new and simple analytical method for glycarbylamide (GB) based on the formation of nickel chelate was developed. The proposed method is as follows: sample solution is mixed with 0.08 mmol/L nickel nitrate in 0.2 mol/L carbonate buffer (pH 9.0)
Daria Pingot et al.
Medycyna pracy, 64(2), 259-271 (2013-07-09)
Acrylamide is a synthetic chemical compound commonly used in many branches of industry. It is mainly used in the synthesis of polyacrylamides, which are widely employed in plastics, paints, varnishes, adhesives and mortars production. Acrylamide is also applied in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service