Skip to Content
Merck
All Photos(2)

Key Documents

32431

Supelco

Furilazole

PESTANAL®, analytical standard

Synonym(s):

(RS)-3-Dichloroacetyl-5-(2-furanyl)-2,2-dimethyl-1,3-oxazolidine

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€283.00

€283.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€283.00

About This Item

Empirical Formula (Hill Notation):
C11H13Cl2NO3
CAS Number:
Molecular Weight:
278.13
Beilstein:
11343170
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€283.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)OC(CN1C(=O)C(Cl)Cl)c2ccco2

InChI

1S/C11H13Cl2NO3/c1-11(2)14(10(15)9(12)13)6-8(17-11)7-4-3-5-16-7/h3-5,8-9H,6H2,1-2H3

InChI key

MCNOFYBITGAAGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Furilazole is classified under the dichloroacetamide group of herbicide safeners.[1]

Application

Furilazole may be used as an analytical reference standard for the determination of the analyte in:
  • Vegetables[2][3] and a fruit samples[2] by solid-phase extraction (SPE) and gas chromatography/multiphoton ionization/time-of-flight mass spectrometry (GC/MPI/TOFMS) as well as GC coupled to electron ionization (EI) MS.
  • Food samples by GC/MS with multiple reaction monitoring (MRM) detection.[4]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility, partitioning, oxidation and photodegradation of dichloroacetamide herbicide safeners, benoxacor and furilazole.
Acharya SP and Weidhaas J
Chemosphere, 211, 1018-1024 (2018)
Ionization of pesticides using a far-ultraviolet femtosecond laser in gas chromatography/time-of-flight mass spectrometry.
Hashiguchi Y, et al.
Analytical and Bioanalytical Chemistry, 405(22), 7053-7059 (2013)
Compensation of matrix effects in gas chromatography-mass spectrometry analysis of pesticides using a combination of matrix matching and multiple isotopically labeled internal standards.
Tsuchiyama T, et al.
Journal of Chromatography A, 1524(22), 233-245 (2017)
Analysis of pesticides by gas chromatography/multiphoton ionization/mass spectrometry using a femtosecond laser.
Li A, et al.
Analytica Chimica Acta, 701(1), 52-59 (2011)
Md Shiful Islam et al.
Gene, 639, 149-162 (2017-10-11)
Glutathione S-transferases (GSTs) are ubiquitous enzymes which play versatile functions including cellular detoxification and stress tolerance. In this study, a comprehensive genome-wide identification of GST gene family was carried out in potato (Solanum tuberosum L.). The result demonstrated the presence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service