Skip to Content
Merck
All Photos(1)

Key Documents

32430

Supelco

Cafenstrole

PESTANAL®, analytical standard

Synonym(s):

N,N-Diethyl-3-mesitylsulfonyl-1H-1,2,4-triazole-1-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N4O3S
CAS Number:
Molecular Weight:
350.44
Beilstein:
8432754
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCN(CC)C(=O)n1cnc(n1)S(=O)(=O)c2c(C)cc(C)cc2C

InChI

1S/C16H22N4O3S/c1-6-19(7-2)16(21)20-10-17-15(18-20)24(22,23)14-12(4)8-11(3)9-13(14)5/h8-10H,6-7H2,1-5H3

InChI key

HFEJHAAIJZXXRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Abd El-Aty et al.
Biomedical chromatography : BMC, 22(3), 306-315 (2007-12-07)
The present work reports the extraction and clean-up procedures, as well as the chromatographic conditions developed, for the determination of cafenstrole and its metabolite (CHM-03) residues in brown rice grains and rice straw using HPLC-UV detection. The method makes use
Takashi Nobusawa et al.
Genes to cells : devoted to molecular & cellular mechanisms, 17(8), 709-719 (2012-06-28)
Recent studies have showed the essential mechanisms for plastid division that have bacterial orthologues, such as FtsZ and Min system proteins; however, causal factors regulating plastid division in plant cells are poorly understood. Here, we show that plastid division is
Olga Ratai et al.
Frontiers in molecular neuroscience, 12, 78-78 (2019-04-20)
Neuronal calcium sensor-1 (NCS-1) knockout (KO) in mice (NCS-1-/- mice) evokes behavioral phenotypes ranging from learning deficits to avolition and depressive-like behaviors. Here, we showed that with the onset of adulthood NCS-1-/- mice gain considerable weight. Adult NCS-1-/- mice are
H Takahashi et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 56(9-10), 781-786 (2001-11-29)
The rice herbicide cafenstrole and its analogs inhibited the incorporation of [1-(14)C]-oleate and (2-(14)C]-malonate into very-long-chain fatty acids (VLCFAs), using Scenedesmus cells and leek microsomes from Allium porrum. Although the precise mode of interaction of cafenstrole at the molecular level
Heather D VanGuilder Starkey et al.
The European journal of neuroscience, 37(10), 1643-1658 (2013-02-27)
Myelin-associated inhibitor/NgR1 signaling has important roles in modulation of synaptic plasticity, with demonstrated effects on cognitive function. We have previously demonstrated that NgR1 and its ligands are upregulated in the hippocampus of aged rats with impaired spatial learning and memory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service