Skip to Content
Merck
All Photos(2)

Key Documents

32072

Supelco

Simeton

PESTANAL®, analytical standard

Synonym(s):

2,4-Bis(ethylamino)-6-methoxy-s-triazine, N2,N4-Diethyl-6-methoxy-1,3,5-triazine-2,4-diamine, NSC 163050, Simetone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15N5O
CAS Number:
Molecular Weight:
197.24
Beilstein:
11608
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(NCC)nc(OC)n1

InChI

1S/C8H15N5O/c1-4-9-6-11-7(10-5-2)13-8(12-6)14-3/h4-5H2,1-3H3,(H2,9,10,11,12,13)

InChI key

HKAMKLBXTLTVCN-UHFFFAOYSA-N

General description

Simeton is an alkoxytriazine, which is mostly used as pesticide.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Simeton has been used as internal standard in ESI-LC/MS method for quantifying environmental
contaminants and in LC-MS/MS analysis for determination of pesticide residue from constructed wetlands.3

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purna K Boruah et al.
Chemosphere, 268, 129328-129328 (2020-12-29)
A facile and an eco-friendly reduction and functionalization of reduced graphene oxide (rGO) sheets is carried out using dopamine and decorated with magnetic Fe3O4 nanoparticles with an average size of 12 nm by a simple co-precipitation method which is established as
Wenjing Deng et al.
Ecotoxicology and environmental safety, 125, 121-127 (2015-12-22)
The occurrence and distribution of six typical antibiotics in the main rivers of Hong Kong were investigated using high-performance liquid chromatography electrospray ionization tandem mass spectrometry (HPLC-ES-MS/MS). The results revealed that the antibiotics were widely distributed in the area studied.
Developments in the mechanism of photodegradation of triazine-based pesticides.
Canle, L. M., M. I. Fernandez, and J. A. Santaballa.
Journal of Physical Organic Chemistry, 18.2, 148-155 (2005)
Development of primary standards for mass spectrometry to increase accuracy in quantifying environmental contaminants.
Oates RP et.al.
Journal of Chromatography A, 1506, 134-137 (2017)
Jie Hou et al.
Environmental science and pollution research international, 23(2), 1722-1731 (2015-09-24)
Several parameters of the method, solid-phase extraction (SPE)-ultra-performance liquid chromatography (UPLC)-tandem mass spectrometry (MS/MS), were optimized to investigate the presence and partitioning of 18 antibiotics (including sulfonamides, tetracyclines, quinolones, macrolides, and β-lactams) during various processing stages at two typical pharmaceutical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service