Skip to Content
Merck
All Photos(1)

Key Documents

320528

Sigma-Aldrich

Sulfuryl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SO2Cl2
CAS Number:
Molecular Weight:
134.97
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor density

4.7 (vs air)

vapor pressure

100 mmHg ( 17.8 °C)
105 mmHg ( 20 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.443 (lit.)

bp

68-70 °C (lit.)

mp

−54 °C (lit.)

density

1.665 g/mL at 20 °C (lit.)

SMILES string

ClS(Cl)(=O)=O

InChI

1S/Cl2O2S/c1-5(2,3)4

InChI key

YBBRCQOCSYXUOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derek R Boyd et al.
Organic & biomolecular chemistry, 12(13), 2128-2136 (2014-02-27)
Monocyclic allylic cis-1,2-diols reacted with sulfuryl chloride at 0 °C in a regio- and stereo-selective manner to give 2-chloro-1-sulfochloridates, which were hydrolysed to yield the corresponding trans-1,2-chlorohydrins. At -78 °C, with very slow addition of sulfuryl chloride, cyclic sulfates were
Chlorinations with sulfuryl chloride. I. The peroxide-catalyzed chlorination of hydrocarbons.
Kharasch MS and Brown HC.
Journal of the American Chemical Society, 61(8), 2142-2150 (1939)
The chlorination of active hydrogen compounds with sulfuryl chloride. I. Ketones.
Wyman DP and Kaufman PR.
The Journal of Organic Chemistry, 29(7), 1956-1960 (1964)
Chlorination of Dimethyl Sulfide and Some of Its Derivatives with Sulfuryl Chloride and Thionyl Chloride1.
Truce WE, et al.
Journal of the American Chemical Society, 74(14), 3594-3599 (1952)
Application of Sulfuryl Chloride for the Quick Construction of β-Chlorotetrahydrofuran Derivatives from Homoallylic Alcohols under Mild Conditions.
Zeng X, et al.
Synthesis, 45(17), 2391-2396 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service