Skip to Content
Merck
All Photos(1)

Key Documents

31761

Supelco

Trietazine

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€222.00

€222.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€222.00

About This Item

Empirical Formula (Hill Notation):
C9H16ClN5
CAS Number:
Molecular Weight:
229.71
Beilstein:
16028
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€222.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(Cl)nc(n1)N(CC)CC

InChI

1S/C9H16ClN5/c1-4-11-8-12-7(10)13-9(14-8)15(5-2)6-3/h4-6H2,1-3H3,(H,11,12,13,14)

InChI key

HFBWPRKWDIRYNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trietazine is a triazine herbicide primarily applied as pre- and post- emergent chemical for the control of annual and perennial grass and broad-leaved weeds.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Trietazine may be used as a herbicide reference standard for the determination of the analyte:
  • In milk samples based on microwave-assisted ionic liquid microextraction (MAILME) coupled with high-performance liquid chromatography (HPLC).[2]
  • In traditional Chinese medicines (TCMs) by liquid chromatography combined with ultra fast electrospray ionization tandem mass spectrometry (UFLC-ESI-MS/MS) method.[3]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Süleyman Aşır et al.
Electrophoresis, 36(23), 2888-2895 (2015-08-08)
Trietazine was selectively separated from aqueous solution containing the competitor molecule cyanazine, which is similar in size and shape to the template molecule. Structural features of the molecularly imprinted column were figured out by SEM. The influence of the mobile-phase
Determination of phenylurea and triazine herbicides in milk by microwave assisted ionic liquid microextraction high-performance liquid chromatography.
Gao S, et al.
Talanta, 82(4), 1371-1377 (2010)
E L Balabanova et al.
Gigiena i sanitariia, (8)(8), 50-52 (1990-08-01)
The study was undertaken to analyze the capacity of organic active chlorotriazine pigments to make contact with blood serum proteins. It was established that pigment toxicity was inversely correlated to the capacity of making contact with blood serum proteins (r
Determination of triazine herbicides in environmental samples.
Dean JR, et al.
Journal of Chromatography A, 733(1-2), 295-335 (1996)
Sung-Ben Huang et al.
Journal of agricultural and food chemistry, 51(25), 7252-7258 (2003-12-04)
A method is reported for the determination of atrazine and its dealkylated chlorotriazine metabolites in ground, surface, and deionized water. Water samples are adjusted to pH 3-4 prior to loading onto two SPE cartridges in series: C-18 and C-18/cation exchange

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service