Skip to Content
Merck
All Photos(1)

Documents

31673

Supelco

4,4′-Methylene-bis(2-chloroaniline)

analytical standard

Synonym(s):

2,2′-Dichloro-4,4′-methylenedianiline, 4,4′-Diamino-3,3′-dichlorodiphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[C6H3(Cl)NH2]2
CAS Number:
Molecular Weight:
267.15
Beilstein:
1882318
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

202-214 °C/0.3 mmHg (lit.)

mp

102-107 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Nc1ccc(Cc2ccc(N)c(Cl)c2)cc1Cl

InChI

1S/C13H12Cl2N2/c14-10-6-8(1-3-12(10)16)5-9-2-4-13(17)11(15)7-9/h1-4,6-7H,5,16-17H2

InChI key

IBOFVQJTBBUKMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Luukkanen et al.
Pharmacology & toxicology, 80(3), 152-158 (1997-03-01)
Rats were treated with acetone, pyrazole, phenobarbital, 4,4'-methylenebis-(2-chloroaniline) (MOCA), 3-methylcholanthrene, creosote oil, or a mixture of polychlorinated biphenyls (Aroclor 1254) to study the inducibility and enzyme kinetics of UDP-glucuronosyltransferases towards 1-hydroxypyrene, which is a human metabolite and a urinary biomarker
M Schuler et al.
Mutation research, 392(1-2), 81-95 (1997-08-01)
The in vitro micronucleus assay in conjunction with CREST-staining and fluorescence in situ hybridization (FISH) with centromere-specific DNA probes is being increasingly utilized for the detection of clastogenic and aneuploidy-inducing agents. Although potentially powerful techniques, both methods have unique characteristics
Hong-I Chen et al.
Journal of occupational health, 49(5), 389-398 (2007-10-24)
Oxidative DNA damage may play an important role in the human carcinogenic process. Recently, we reported a case of bladder cancer among 4, 4'-methylenebis (2-chloroaniline) (MBOCA)-exposed workers. By measuring the plasma level of 8-hydroxydeoxyguanosine (8-OHdG), we investigated the association between
4,4'-Methylenebis(2-chloroaniline).
Report on carcinogens : carcinogen profiles, 11, III167-III168 (2004-01-01)
E B Murray et al.
Reviews on environmental health, 20(3), 163-176 (2005-12-13)
Cytogenetic end-points used to estimate risk of genotoxic events in workers include the measurement of micronuclei (MN) in exfoliated cells, lymphocytes, and other tissues. Micronuclei are chromatin-containing bodies outside the cell nucleus resulting from contaminant-induced DNA damage. A review of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service