Skip to Content
Merck
All Photos(1)

Key Documents

31650-M

Sigma-Aldrich

Zinc chloride

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€55.40
1 KG
€182.00

€55.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€55.40
1 KG
€182.00

About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
crystalline powder

€55.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor pressure

1 mmHg ( 428 °C)

Assay

≥98%

form

crystalline powder

impurities

≤0.002% total nitrogen (N)
≤0.005% insoluble in HCl
≤1.2% oxide chloride (as ZnO)

pH

5 (20 °C, 100 g/L)

mp

293 °C (lit.)

anion traces

sulfate (SO42-): ≤20 mg/kg

cation traces

Ca: ≤0.001%
Cd: ≤5 mg/kg
Cu: ≤0.001%
Fe: ≤0.0005%
K: ≤0.02%
Mg: ≤0.001%
Na: ≤0.05%
Pb: ≤0.001%

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc chloride in combination with 1-ethyl-3-methylimidazolium chloride comprises a molten salt, which is employed in the electrodeposition studies of zinc.[1]

Application

Zinc Chloride may be employed as a catalyst in Knoevenagel condensation of carbonyl compounds with active methylene group containing reagents.[2] It may be used in the preparation of porous carbon nanofibers, useful in the fabrication of efficient electrodes for supercapacitors.[3] It may be used as a catalyst in the preparation of poly(propylene fumarate).[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Albert K Shung et al.
Journal of biomaterials science. Polymer edition, 13(1), 95-108 (2002-05-11)
Diethyl fumarate and propylene glycol were reacted in the presence of a zinc chloride catalyst to synthesize poly(propylene fumarate) (PPF) over a period of 12 hours. The kinetics of the transesterification polymerization at 130 degrees C, 150 degrees C, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service