Skip to Content
Merck
All Photos(2)

Key Documents

31562

Supelco

Disulfoton-sulfoxide

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€92.40

€92.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€92.40

About This Item

Empirical Formula (Hill Notation):
C8H19O3PS3
CAS Number:
Molecular Weight:
290.40
Beilstein:
1712528
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€92.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)SCCS(=O)CC

InChI

1S/C8H19O3PS3/c1-4-10-12(13,11-5-2)14-7-8-15(9)6-3/h4-8H2,1-3H3

InChI key

UPUGLJYNCXXUQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disulfoton-sulfoxide is one of the oxidative metabolites of disulfoton, an organophosphorus insecticide used against aphids, mites, and thrips.[1]

Application

Disulfoton-sulfoxide may be used as a reference standard for the determination of the analyte:
  • In human biological samples based on QuEChERS (Quick, Easy, Cheap, Effective, Rugged, and Safe) extraction and liquid chromatography–tandem mass spectrometry.[1]
  • In cereal-based baby foods and wheat flour using hollow-fiber liquid-phase microextraction prior to gas chromatography with nitrogen phosphorus detection.[2]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of organophosphorus pesticides and metabolites in cereal-based baby foods and wheat flour by means of ultrasound-assisted extraction and hollow-fiber liquid-phase microextraction prior to gas chromatography with nitrogen phosphorus detection.
Gonzalez-Curbelo MA, et al.
Journal of Chromatography A, 1313(6), 166-174 (2013)
Rapid determination of disulfoton and its oxidative metabolites in human whole blood and urine using QuEChERS extraction and liquid chromatography-tandem mass spectrometry.
Usui K, et al.
Legal medicine, 14(6), 309-316 (2012)
Yasuhisa Fujino et al.
Chudoku kenkyu : Chudoku Kenkyukai jun kikanshi = The Japanese journal of toxicology, 17(3), 273-277 (2004-10-22)
We encountered three cases of organophosphate poisoning treated with pralidoxime iodide (PAM) and whole-bowel irrigation without atropine sulfate. All patients recovered without persistence or recurrence of toxic symptoms and without any somatic after effects. In case 1, a 48-year-old woman
B Dementi
Journal of applied toxicology : JAT, 14(2), 119-129 (1994-03-01)
Many publications, primarily of work performed in Japan, report findings in human populations of an increased incidence of myopia and of a more advanced visual disease syndrome (Saku disease), which reportedly correlated with increasing use of organophosphate pesticides in agriculture.
Jishi Wang et al.
Journal of chromatography. A, 1521, 10-18 (2017-09-25)
In this research, the manual two-step QuEChERS approach has been streamlined and automated into a one-step method using a cleanup tube fitted within an extraction tube. A novel automatic QuEChERS combination have been developed to simplify the QuEChERS procedures and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service