Skip to Content
Merck
All Photos(1)

Documents

309125

Sigma-Aldrich

Barium peroxide

−325 mesh, 95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaO2
CAS Number:
Molecular Weight:
169.33
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38110504
PubChem Substance ID:

Assay

95%

reaction suitability

reagent type: oxidant

particle size

−325 mesh

SMILES string

[Ba++].[O-][O-]

InChI

1S/Ba.O2/c;1-2/q+2;-2

InChI key

ZJRXSAYFZMGQFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service