Skip to Content
Merck
All Photos(1)

Key Documents

30709-M

Sigma-Aldrich

Nitric acid

puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€47.40
2.5 L
€103.00

€47.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€47.40
2.5 L
€103.00

About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

€47.40


Please contact Customer Service for Availability

Request a Bulk Order

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Quality Level

vapor pressure

8 mmHg ( 20 °C)

description

for determinations with dithizone

grade

puriss. p.a.

Assay

≥65%

form

liquid

ign. residue

≤0.0005% (as SO4)

bp

120.5 °C (lit.)

density

1.37-1.41 g/mL at 20 °C (lit.)

anion traces

chloride (Cl-): ≤0.00005%
phosphate (PO43-): ≤0.00005%
sulfate (SO42-): ≤0.00005%

cation traces

Ag: ≤0.000001%
Al: ≤0.000005%
As: ≤0.000001%
Ba: ≤0.000001%
Be: ≤0.000001%
Bi: ≤0.00001%
Ca: ≤0.00001%
Cd: ≤0.000001%
Co: ≤0.000001%
Cr: ≤0.000002%
Cu: ≤0.000001%
Fe: ≤0.00001%
Ge: ≤0.000005%
Hg: ≤0.000001%
K: ≤0.000005%
Li: ≤0.000001%
Mg: ≤0.00001%
Mn: ≤0.000001%
Mo: ≤0.000002%
Na: ≤0.00005%
Ni: ≤0.000002%
Pb: ≤0.000001%
Sr: ≤0.000001%
Ti: ≤0.00001%
Tl: ≤0.000005%
V: ≤0.000001%
Zn: ≤0.000005%
Zr: ≤0.00001%

SMILES string

O[N+]([O-])=O

suitability

passes test for suitability for det. with dithizone

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitric acid (HNO3) is a highly corrosive strong mineral acid. 10%(v/v) nitric acid solution is useful for cleaning laboratory glasswares.[1] Reaction of HNO3 with various original and synthetic mineral dust/mineral oxide surfaces was studied in a low-pressure Knudsen reactor operating at 298K.[2]

Application

Nitric acid (HNO3) may be used for the removal of transition metal catalyst from the single-walled carbon nanotubes (SWNTs) during the purification of SWNTs.[3] It may be used in the trace determinations of Pb and Cd in natural waters by flame atomic absorption spectrometry.[1]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Carasek et al.
Talanta, 56(1), 185-191 (2008-10-31)
The need for highly reliable methods for the determination of trace elements has been recognised in analytical chemistry and environmental science. A method for the trace analysis of Pb and Cd in natural waters is described. In a preconcentration step

Questions

  1. Hi, I cannot find the type of package - glass, FEP ... Can you clarify that? Can this type of acid be used for Trace elemental analysis?

    1 answer
    1. This product is packaged in amber glass bottles. See the 'PROPERTIES' section above, under anion and cation traces, to review the elemental testing and specifications for this material. Yes, this product is suitable for use in trace metal analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service