30512
Decahydro-2-naphthol
purum, ≥98.0% (GC)
Synonym(s):
2-Decalol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
vapor density
>1 (vs air)
grade
purum
Assay
≥98.0% (GC)
refractive index
n20/D 1.5 (lit.)
bp
109 °C/14 mmHg (lit.)
density
0.996 g/mL at 25 °C (lit.)
SMILES string
OC1CCC2CCCCC2C1
InChI
1S/C10H18O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h8-11H,1-7H2
InChI key
UPMAOXLCTXPPAG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Components
mixture of isomers (~70% 2-decalol)
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service