Skip to Content
Merck
All Photos(1)

Documents

30230

Sigma-Aldrich

L-Cystine bisallyl ester di(p-toluenesulfonate) salt

≥95.0% (sum of enantiomers, TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2OCOCH(NH2)CH2SSCH2CH(NH2)COOCH2CH=CH2.2C7H8O3S
CAS Number:
Molecular Weight:
664.83
Beilstein:
7966375
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95.0% (sum of enantiomers, TLC)

optical activity

[α]20/D −11.5±1°, c = 3% in methanol

mp

175-180 °C

SMILES string

Cc1ccc(cc1)S(O)(=O)=O.Cc2ccc(cc2)S(O)(=O)=O.N[C@@H](CSSC[C@H](N)C(=O)OCC=C)C(=O)OCC=C

Other Notes

Used in peptide and N-glycopeptide synthesis; the ester is selectively deblocked with (Ph3P)3RhCl

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Waldmann et al.
Liebigs Ann. Chem., 1712-1712 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service