Skip to Content
Merck
All Photos(1)

Key Documents

30172

Supelco

Arachidonyl-L-carnitine

analytical standard

Synonym(s):

C20:4(n-6) carnitine, (2R)-3-Carboxy-N,N,N-trimethyl-2-[[(5Z,8Z,11Z,14Z)-1-oxo-5,8,11,14-eicosatetraen-1-yl]oxy]-1-propanaminium inner salt, Arachidonoylcarnitine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€180.00

€180.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€180.00

About This Item

Empirical Formula (Hill Notation):
C27H45NO4
CAS Number:
Molecular Weight:
447.65
MDL number:
UNSPSC Code:
41116107
NACRES:
NB.21

€180.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (LC/MS)

optical activity

[α]/D -13±3°, c = 0.1 in methanol

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

[O-]C(C([H])([H])[C@](OC(C([H])([H])C([H])([H])C([H])([H])/C([H])=C([H])\C([H])([H])/C([H])=C([H])\C([H])([H])/C([H])=C([H])\C([H])([H])/C([H])=C([H])\C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])[H])=O)([H])C([H])([H])[N+](C([H])([H])[H])(C([H]

General description

Arachidonyl-L-carnitine belongs to the class of long-chain acylcarnitines,[1] a wide range of metabolites, derived from their cognate acyl-CoAs. They serve as good biomarkers for perturbations of a multitude of distinct metabolic pathways.[2]

Application

Arachidonyl-L-carnitine may be used as an analytical reference standard for the quantification of the analyte in cultured cells using hydrophilic interaction liquid chromatography (HILIC) and liquid chromatography–electrospray ionization tandem mass spectrometry (LC–ESI-MS/MS).[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new LC?ESI-MS/MS method to measure long-chain acylcarnitine levels in cultured cells
Jauregui O, et al.
Analytica Chimica Acta, 599(1), 1-6 (2007)
Presence of arachidonoyl-carnitine is associated with adverse cardiometabolic responses in hypertensive patients treated with atenolol
Weng L, et al.
Metabolomics, 12(10), 160-160 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service