Skip to Content
Merck
All Photos(1)

Key Documents

28078

Supelco

9,11-Didehydrooestriol

≥97.0% (HPLC)

Synonym(s):

Estra-1,3,5,9(11)-tetraene-3,16α,17β-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O3
CAS Number:
Molecular Weight:
286.37
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97.0% (HPLC)

SMILES string

[H][C@@]12CCc3cc(O)ccc3C1=CC[C@]4(C)[C@@H](O)[C@H](O)C[C@@]24[H]

InChI

1S/C18H22O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5-6,8,14-17,19-21H,2,4,7,9H2,1H3/t14-,15+,16-,17+,18+/m1/s1

InChI key

ZWHFPHWLYUMTGY-WKULXVSPSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service