Key Documents
27383
Supelco
Hamilton® plunger assemblies with tips for series 1000, 1700, and 1800
for use with Hamilton 1801, N, RN, pkg of 1 ea
About This Item
UNSPSC Code:
41122001
material
PTFE plunger tip
size
10 μL
feature
1801N, 1801RN
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 32193
compatibility
for use with Hamilton 1801, N, RN
General description
For field repair of Hamilton Series 1000, 1700 and 1800 syringes.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)