Skip to Content
Merck
All Photos(1)

Key Documents

26423

Sigma-Aldrich

Zinc stearate

purum, 10-12% Zn basis

Synonym(s):

Stearic acid zinc salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€40.40
10 KG
€279.00

€40.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
€40.40
10 KG
€279.00

About This Item

Linear Formula:
[CH3(CH2)16COO]2Zn
CAS Number:
Molecular Weight:
632.33
Beilstein:
3919706
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€40.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

grade

purum

autoignition temp.

1454 °F

composition

mixture consisting mainly of palmitate (C16) and stearate (C18) salts, 90%
stearate salt : Minimum, ~40%

reaction suitability

core: zinc
reagent type: catalyst

concentration

10-12% Zn

impurities

≤3.5% free acid (as stearic acid)

ign. residue

12-15%

loss

≤1% loss on drying, 105 °C

mp

128-130 °C (lit.)

solubility

alcohol: insoluble(lit.)
benzene: soluble(lit.)
diethyl ether: insoluble(lit.)
water: insoluble(lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Zn]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Zn/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

XOOUIPVCVHRTMJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

The effect of zinc stearate on mechanical properties and on thermal stability of plastigels obtained from polyvinylchloride and dioctylphthalate plastisols was studied[1]. Zinc stearate decreases the melt viscosity of the ionomeric blend of maleated ethylene-propylene-diene monomers and maleated high-density polyethylene[2].

Application

Zinc stearate has been used as reinforcing filler and as a plasticizer for the maleated ethylene-propylene-diene rubber[3].

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of zinc stearate on melt-processable ionomeric blends based on zinc salts of maleated high-density polyethylene and maleated EPDM rubber.
Antony P and De SK.
Polymer, 40(6), 1487-1493 (1999)
Ionic thermoplastic elastomer based on maleated EPDM rubber. I. Effect of zinc stearate.
Datta S, et al.
Journal of Applied Polymer Science, 61(1), 177-186 (1996)
Effect of zinc stearate and/or epoxidized soybean oil on gelation and thermal stability of PVC-DOP plastigels.
Baltacioglu H and Balkose D.
Journal of Applied Polymer Science, 74(10), 2488-2498 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service