Skip to Content
Merck
All Photos(1)

Key Documents

262560

Sigma-Aldrich

Mineral Spirits

odorless, contains ≥10 ppm BHT

Synonym(s):

Ligroin, Petroleum spirits

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
920-901-0
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21
bp:
179-210 °C
vapor pressure:
2 psi ( 21.1 °C)
Product 262560 is not currently sold in your country. Contact Technical Service

vapor density

>1 (vs air)

Quality Level

vapor pressure

2 psi ( 21.1 °C)

form

liquid

contains

≥10 ppm BHT

refractive index

n20/D 1.424

bp

179-210 °C

density

0.752 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Mineral spirits is a petrol-based aliphatic hydrocarbon solvent commonly used as degreaser.[1]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Airborne concentrations of benzene and mineral spirits (stoddard solvent) during cleaning of a locomotive generator and traction motor
Madl AK & Paustenbach DJ
Environmental Health : A Global Access Science Source, 65(23), 1965-1979 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service