Skip to Content
Merck
All Photos(1)

Key Documents

25668

Sigma-Aldrich

Chloroform – isoamyl alcohol mixture

BioUltra, for molecular biology, 49:1, ≥99.5% (chloroform + isoamyl alcohol, GC)

Synonym(s):

Chloroform:Isoamyl Alcohol Solvent Mix

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€84.30
500 ML
€317.00

€84.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€84.30
500 ML
€317.00

About This Item

MDL number:
UNSPSC Code:
12352401
PubChem Substance ID:
NACRES:
NA.25
grade:
Molecular Biology
for molecular biology
technique(s):
GC/GC: suitable

€84.30


Please contact Customer Service for Availability

Request a Bulk Order

grade

Molecular Biology
for molecular biology

Quality Level

product line

BioUltra

Assay

≥99.5% (chloroform + isoamyl alcohol, GC)

form

liquid

technique(s)

GC/GC: suitable

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.001% free alkali (as NH3)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.1
λ: 280 nm Amax: 0.07

SMILES string

ClC(Cl)Cl.CC(C)CCO

InChI

1S/C5H12O.CHCl3/c1-5(2)3-4-6;2-1(3)4/h5-6H,3-4H2,1-2H3;1H

InChI key

BKHZIBWEHPHYAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chloroform – isoamyl alcohol mixture is a deproteinizing reagent used in conjunction with phenol for improving the efficiency of RNA and DNA (nucleic acid) extractions.

Components

mixture of chloroform and isoamyl alcohol in the ratio of 49:1 (v/v)

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.M. Wallace
Methods in Enzymology, 152, 35-35 (1987)
Purification of RNA by SDS solubilization and phenol extraction.
Rio DC, Ares M Jr, Hannon GJ, Nilsen TW.
Cold Spring Harbor Protocols, 2010, 5439-5439 (2010)
Bacterial RNA isolation.
Ares M.
Cold Spring Harbor Protocols, 1012, 1024-1027 (2012)
Deanna L Davis et al.
Journal of lipid research, 61(4), 505-522 (2020-02-12)
Myelin is a unique lipid-rich membrane structure that accelerates neurotransmission and supports neuronal function. Sphingolipids are critical myelin components. Yet sphingolipid content and synthesis have not been well characterized in oligodendrocytes, the myelin-producing cells of the CNS. Here, using quantitative
Marion Poirier et al.
ChemMedChem, 14(2), 224-236 (2018-12-07)
By screening a focused library of kinase inhibitor analogues in a phenotypic co-culture assay for angiogenesis inhibition, we identified an aminotriazine that acts as a cytostatic nanomolar inhibitor. However, this aminotriazine was found to be completely inactive in a whole-kinome

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service