Key Documents
24582
Hamilton® needles for RN syringes, cone tip
volume 5-10 μL, ASRN (Autosampler Removable Needle), pkg of 6 ea
About This Item
UNSPSC Code:
41122001
material
stainless steel
needle size
(Pt. Style 1 (cone tip))
feature
ASRN (Autosampler Removable Needle)
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 7785-01
needle L
43 mm
volume
5-10 μL
compatibility
for use with Hamilton 5μL-10μL syringe
for use with Hamilton Hamilton Gastight SyringeModel 175ASRN
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)