Skip to Content
Merck
All Photos(1)

Key Documents

24523

Hamilton® syringes for PerkinElmer® GC Autosystem and Clarus 500

75 ASN/PE GC, volume 5 μL, needle size 23s ga (blunt tip), needle L 70 mm, needle type, LTN (fixed)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€109.00

€109.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€109.00

About This Item

UNSPSC Code:
41122001

€109.00


Please contact Customer Service for Availability

Request a Bulk Order

material

blunt tip stainless steel needle (point style 3)

needle size

23s ga (blunt tip)

feature

needle type LTN (fixed)

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 88035

needle L

70 mm

volume

5 μL

compatibility

for use with PerkinElmer GC Autosystem

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.
PerkinElmer is a registered trademark of PerkinElmer Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wolf, Don P.
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192-192 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service