Skip to Content
Merck
All Photos(1)

Documents

224863

Sigma-Aldrich

Potassium chlorate

reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KClO3
CAS Number:
Molecular Weight:
122.55
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

reagent grade

Assay

98%

mp

356 °C (lit.)

SMILES string

[K+].[O-]Cl(=O)=O

InChI

1S/ClHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

VKJKEPKFPUWCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola González Audino et al.
Pest management science, 58(2), 183-189 (2002-02-21)
The stability to heart of cis-permethrin and beta-cypermethrin in the solid phase was studied and the decomposition products identified. Samples heated at 210 degrees C in an oven in the dark showed that, in the absence of potassium chlorate (the
Kazutomo Ishi et al.
Bioscience, biotechnology, and biochemistry, 69(12), 2463-2465 (2005-12-27)
The nitrate reductase gene (niaD) is the most frequently utilized as a selectable marker for homologous integration at the niaD locus of Aspergillus oryzae. In this study we developed a method for curing of the niaD-based plasmid integrated on the
Sang Jin Lee et al.
Journal of biomaterials science. Polymer edition, 13(2), 197-212 (2002-05-23)
It has been recognized that adhesion and proliferation of cells on biodegradable polymers such as poly(lactic acid) (PLA), poly(glycolic acid) (PGA), and poly(lactide-co-glycolide) (PLGA) depend on the surface properties. The chloric acid (CA) treatment of these films was developed to
Md Monirul Islam et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 26(1), 13-18 (2010-01-13)
Interdigitated array electrodes with surface-enhanced Raman scattering (SERS) functionality for in situ qualitative and quantitative analysis of electroactive species are demonstrated. Gold nanostructured interdigitated array electrodes (NIDAEs) were fabricated by electron beam lithography, and used for an electrochemical SERS study
F Khan et al.
Biochimica et biophysica acta, 1481(2), 229-236 (2000-10-06)
Characterization of conformational transition and folding intermediates is central to the study of protein folding. We studied the effect of various alcohols (trifluoroethanol (TFE), butanol, propanol, ethanol and methanol) and salts (K(3)FeCN(6), Na(2)SO(4), KClO(4) and KCl) on the acid-induced state

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service