Skip to Content
Merck
All Photos(1)

Key Documents

222542

Sigma-Aldrich

N-(Diphenylmethylene)glycine ethyl ester

98%

Synonym(s):

Ethyl (diphenylmethylenamino)acetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€189.00
25 G
€694.00

€189.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€189.00
25 G
€694.00

About This Item

Linear Formula:
(C6H5)2C=NCH2CO2C2H5
CAS Number:
Molecular Weight:
267.32
Beilstein:
1979922
MDL number:
UNSPSC Code:
12171500
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.47

€189.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder, crystals or chunks

impurities

<2% benzophenone

mp

51-53 °C (lit.)

solubility

ethanol: 50 mg/mL, clear to very slightly hazy, colorless to faint yellow or tan

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

CCOC(=O)C\N=C(/c1ccccc1)c2ccccc2

InChI

1S/C17H17NO2/c1-2-20-16(19)13-18-17(14-9-5-3-6-10-14)15-11-7-4-8-12-15/h3-12H,2,13H2,1H3

InChI key

QUGJYNGNUBHTNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Schiff base which can be converted to amino alcohols[1][2] and amino esters.[3][4]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 57, 5469-5469 (1992)
Journal of the Chemical Society. Chemical Communications, 682-682 (1991)
Tetrahedron Letters, 34, 8535-8535 (1993)
Tetrahedron Letters, 34, 677-677 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service