Skip to Content
Merck
All Photos(1)

Documents

220752

Supelco

Florisil®

reagent grade, 60-100 mesh

Synonym(s):

Activated magnesium silicate, Magnesium silicate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO3Si
CAS Number:
Molecular Weight:
100.39
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:

grade

reagent grade

particle size

60-100 mesh

transition temp

transition temp 675 °C (activation temperature)

SMILES string

[Mg++].[O-][Si]([O-])=O

InChI

1S/Mg.O3Si/c;1-4(2)3/q+2;-2

InChI key

FKHIFSZMMVMEQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An activated magnesium silicate suitable for use in pesticide-residue analysis.

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

U Avsar et al.
Cellular and molecular biology (Noisy-le-Grand, France), 59 Suppl, OL1835-OL1841 (2013-02-13)
This study evaluated the protective effect of Panax Ginseng (PG) on bone metabolism in an experimental ovariectomy (OVX) model of osteoporosis in which inflammation was induced by subcutaneous magnesium silicate. The groups were: sham control (Group1, SH), sham+inflammation (Group2, SHinf)
Sangram H Patil et al.
Journal of chromatography. A, 1216(12), 2307-2319 (2009-02-14)
A multiresidue method is described for simultaneous estimation of 83 pesticides and 12 dioxin-like polychlorinated biphenyls (PCBs) in red and white wines. The samples (20mL wine, acidified with 20 mL 1% HCl) were extracted with 10 mL ethyl acetate (+20
Li Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(16), 6292-6295 (2013-04-12)
Knowledge of the structural properties of mantle phases is critical for understanding the enigmatic seismic features observed in the Earth's lower mantle down to the core-mantle boundary. However, our knowledge of lower mantle phase equilibria at high pressure (P) and
Lei Zhang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 63(10), 2114-2122 (2011-10-08)
Florisil was employed for the sorption of antimony ions from aqueous solutions. A detailed study of the process was performed by varying the sorption time, pH, and temperature. The sorption was found to be fast, equilibrium was reached within 15
Iyad Rashid et al.
International journal of pharmaceutics, 411(1-2), 18-26 (2011-03-23)
A directly compressible excipient has been developed by co-processing starch with magnesium silicate. The foregoing was achieved either by co-precipitation of magnesium silicate onto different types of starch or by dry granulation of maize starch with magnesium silicate. A variety

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service