Skip to Content
Merck
All Photos(2)

Key Documents

20885

Hamilton® HPLC syringes

1701RNR, volume 10 μL, needle size 22s ga (blunt tip), needle L 51 mm (2 in.)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€69.10

€69.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€69.10

About This Item

UNSPSC Code:
41122001

€69.10


Please contact Customer Service for Availability

Request a Bulk Order

material

blunt tip stainless steel needle (point style 3)
glass body

needle size

22s ga (blunt tip)

feature

blunt tip

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 80065

needle L

51 mm (2 in.)

volume

10 μL

compatibility

for use with Rheodyne
for use with SSI
for use with Valco VISF-2

Looking for similar products? Visit Product Comparison Guide

Application

Hamilton® HPLC syringes may be used for extraction and injection of the sample material.[1]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single drop liquid?liquid?liquid microextraction of methamphetamine and amphetamine in urine
He Y and Kang J-Y
Journal of Chromatography A, 1133, 35-40 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service