Skip to Content
Merck
All Photos(1)

Documents

19355

Supelco

10-Hydroxydecyl butyrate

Selectophore, ≥98.0%

Synonym(s):

ETH 264

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O3
CAS Number:
Molecular Weight:
244.37
Beilstein:
2252705
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

Assay

≥98.0% (GC)
≥98.0%

refractive index

n20/D 1.448

density

0.932 g/mL at 20 °C (lit.)

SMILES string

CCCC(=O)OCCCCCCCCCCO

InChI

1S/C14H28O3/c1-2-11-14(16)17-13-10-8-6-4-3-5-7-9-12-15/h15H,2-13H2,1H3

InChI key

KKIBZIXNDVWPDF-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Plasticizer suitable for polymer membrane matrices, e.g. cadmium selective electrodes; Plasticizer in a bicarbonate-sensitive liquid membrane electrode

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.K. Schneider
Helvetica Chimica Acta, 63, 217-217 (1980)
R.J.J. Funck et al.
Analytical Chemistry, 54, 423-423 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service