Skip to Content
Merck
All Photos(1)

Key Documents

18280

Sigma-Aldrich

3-Bromopyridine

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4BrN
CAS Number:
Molecular Weight:
158.00
Beilstein:
105880
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.571 (lit.)
n20/D 1.571

bp

173 °C (lit.)

density

1.64 g/mL at 25 °C (lit.)

SMILES string

Brc1cccnc1

InChI

1S/C5H4BrN/c6-5-2-1-3-7-4-5/h1-4H

InChI key

NYPYPOZNGOXYSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Bromopyridine was used in the synthesis of cis-1-methyl-1,2,3,3a,4,8b-hexahydropyrrolo[3,2-f]pyrindine (a nicotine analog). It was also used in the preparation of 3-pyridylboronic acid.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongbin Zhai et al.
Organic letters, 4(25), 4385-4386 (2002-12-06)
[reaction: see text] The title compound, 2, has been synthesized in 45% overall yield in six steps from 3-bromopyridine. The hexahydropyrrolo[3,2-f]pyrindine skeleton was constructed from key intermediate 5, via intramolecular azomethine ylide-alkene [3 + 2] cycloaddition. The present work constitutes
Wenjie Li et al.
The Journal of organic chemistry, 67(15), 5394-5397 (2002-07-20)
3-Pyridylboronic acid was prepared in high yield and bulk quantity from 3-bromopyridine via a protocol of lithium-halogen exchange and "in situ quench". This technique was further studied and evaluated on other aryl halides in the preparation of arylboronic acids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service