Skip to Content
Merck
All Photos(1)

Key Documents

17274

Millipore

Thermoacidurans Agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€260.00

€260.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€260.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

€260.00


Please contact Customer Service for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 20 g/L
dextrose, 5 g/L
dipotassium phosphate, 4 g/L
manganese sulfate, 0.2 g/L
proteose peptone, 5 g/L
yeast extract, 5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

5.0±0.2 (25 °C)

application(s)

bioburden testing
food and beverages

microbiology

suitability

nonselective for Bacillus spp.

Application

For isolation and cultivation of Bacillus thermoacidurans from food products.

Preparation Note

Suspend 39.2 g in 1 litre distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 minutes. Cool to 50°C and pour into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service