Skip to Content
Merck
All Photos(1)

Key Documents

16917

Supelco

1,5-Dicaffeoylquinic acid

analytical standard

Synonym(s):

(1S,3R,4R,5R)-1,3-Bis[3-(3,4-dihydroxyphenyl)acryloyloxy]-4,5-dihydroxycyclohexanecarboxylic acid, 1,3-Dicaffeoylquinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24O12
CAS Number:
Molecular Weight:
516.45
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1C[C@@](C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@@H]1O)(OC(=O)\C=C\c3ccc(O)c(O)c3)C(O)=O

InChI

1S/C25H24O12/c26-15-5-1-13(9-17(15)28)3-7-21(31)36-20-12-25(24(34)35,11-19(30)23(20)33)37-22(32)8-4-14-2-6-16(27)18(29)10-14/h1-10,19-20,23,26-30,33H,11-12H2,(H,34,35)/b7-3+,8-4+/t19-,20-,23-,25+/m1/s1

InChI key

YDDUMTOHNYZQPO-PSEXTPKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dicaffeoylquinic acid is naturally occurring polyphenolic compound found in plants like fennel and coffee. It is characterized by two caffeic acid molecules linked via ester bonds to one quinic acid molecule. 1, 5-Dicaffeoylquinic acid (1,5-DCQA) is considered as a potent and nontoxic inhibitor of HIV-1 replication.[1]
In literature, CAS Numbers 19870-46-3 and 30964-13-7 are each described as 1,3-dicaffeoylquinic acid, 1,5-dicaffeoylquinic acid, and cynarin. The IUPAC synonym for this product is provided for clarification.

Application

It may be used as reference standard in LC-MS/MS method for the determination of 1,5-DCQA from human plasma.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Phenolic compound found in artichoke. Antioxidant and choleretic properties. Potential immunosuppressive agent.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: achillea arnica

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruolan Gu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 852(1-2), 85-91 (2007-02-03)
1,5-Dicaffeoylquinic acid (1,5-DCQA), a potent HIV-1 integrase inhibitor, is undergoing an evaluation as a promising novel HIV therapeutic agent. Here, we report a simple, rapid and robust LC-MS/MS method for simultaneous determination of 1,5-DCQA and its two active metabolites, 1-caffeoyl-5-feruoylquinic
Bo Yang et al.
Drug metabolism and disposition: the biological fate of chemicals, 33(7), 930-936 (2005-04-02)
To explore the metabolism of 1,5-dicaffeoylquinic acid (1,5-DCQA) in rats, liquid chromatography-mass spectrometry in parallel to diode-array detection was used for the rapid detection/characterization of the metabolites formed in bile, urine, and plasma of rats following oral administration of 1,5-DCQA
Reham Wagdy et al.
Journal of food science, 84(5), 1012-1022 (2019-04-25)
Dietary phenolics are known for their potent antioxidant and anti-inflammatory activities, making them promising candidates for protection against neuroinflammation and neurodegeneration. Hydroalcohol extract of Egyptian species of Corchorus olitorius L. (Co) leaves was investigated for its neuroprotective effects in a
Daniil N Olennikov et al.
Antioxidants (Basel, Switzerland), 8(8) (2019-08-17)
Fringed sagewort (Artemisia frigida Willd., Compositae family) is a well-known medicinal plant in Asian medical systems. Fifty-nine hydroxycinnamates and flavonoids have been found in A. frigida herbs of Siberian origin by high-performance liquid chromatography with diode array and electrospray triple
Muhammad Mir Showkat et al.
Molecules (Basel, Switzerland), 24(18) (2019-09-13)
Phenolic acids including chlorogenic acids are major polyphenolic compounds found in Jerusalem artichoke (Helianthus tuberosus L.). The plant itself is an emerging biorefinery crop due to the inulin-rich tubers, a bioethanol feedstock, but the aerial parts represent a rich source

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service