Iron phthalocyanine (FePc) is a promising non-precious catalyst for the oxygen reduction reaction (ORR). Unfortunately, FePc with plane-symmetric FeN4 site usually exhibits an unsatisfactory ORR activity due to its poor O2 adsorption and activation. Here, we report an axial Fe-O
Two dimensional (2D)SnO2 nanosheets were synthesized by a substrate-free hydrothermal route using sodium stannate and sodium hydroxide in a mixed solvent of absolute ethanol and deionized water at a lower temperature of 130 °C. The characterization results of the morphology
Manganese dioxide (MnO₂), as a promising green material, has recently attracted considerable attention of researchers from various fields. In this work, a facile method was introduced to prepare binary hybrids by fabricating three-dimensional (3D) zinc hydroxystannate (ZHS) cubes on two-dimensional
An electrochemical immunosensor for ultrasensitive detection of acrylamide (AA) in water and food samples was developed. SnO2-SiC hollow sphere nanochains with high surface area and gold nanoparticles with good electroconductivity were fabricated onto the surface of a glassy carbon electrode