Skip to Content
Merck
All Photos(1)

Key Documents

14288

Sigma-Aldrich

(R)-(+)-2-(Methoxymethoxy)-1-propanol

Synonym(s):

MOM protected (R)-1,2-Propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O3
CAS Number:
Molecular Weight:
120.15
MDL number:

SMILES string

COCO[C@H](C)CO

InChI

1S/C5H12O3/c1-5(3-6)8-4-7-2/h5-6H,3-4H2,1-2H3/t5-/m1/s1

InChI key

YNDXFEKPXUXDQS-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.9 °F

Flash Point(C)

75.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service