Skip to Content
Merck
All Photos(1)

Documents

13613

Sigma-Aldrich

Nickel(II) chloride hexahydrate

puriss., ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiCl2 · 6H2O
CAS Number:
Molecular Weight:
237.69
MDL number:
UNSPSC Code:
12352300

grade

puriss.

Assay

≥97.0%

pH

4-6 (20 °C, 5%)

anion traces

sulfate (SO42-): ≤200 mg/kg

cation traces

As: ≤1 mg/kg
Cd: ≤20 mg/kg
Co: ≤100 mg/kg
Cu: ≤20 mg/kg
Fe: ≤50 mg/kg
Pb: ≤10 mg/kg
Zn: ≤50 mg/kg

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ni]Cl

InChI

1S/2ClH.Ni.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

LAIZPRYFQUWUBN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service