Skip to Content
Merck
All Photos(1)

Key Documents

13418-M

Sigma-Aldrich

Sodium carbonate

puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€47.00
6 X 1 KG
€271.00

€47.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
€47.00
6 X 1 KG
€271.00

About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder

€47.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

grade

puriss.

Assay

99.5-100.5% (calc. to the dried substance)

form

powder

quality

meets analytical specification of Ph. Eur., BP, NF, FCC, E500

impurities

alkali hydroxides and bicarbonates, complies
residual solvents, complies
≤5 ppm heavy metals (as Pb)

loss

≤0.5% loss on drying, 300 °C
<0.5% loss on drying, 105°C, 4hrs

pH

12 (25 °C, 106 g/L)

mp

851 °C (lit.)

anion traces

chloride (Cl-): ≤100 ppm
sulfate (SO42-): ≤50 ppm

cation traces

As: ≤0.0002%
Cu: ≤0.002%
Fe: ≤0.001%
Hg: ≤0.0001%
Pb: ≤0.0002%
Zn: ≤0.002%

suitability

complies for appearance of solution

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service