Skip to Content
Merck
All Photos(1)

Documents

12827

Sigma-Aldrich

Copper(II) chloride dihydrate

technical, ≥98% (complexometric)

Synonym(s):

Cupric chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2 · 2H2O
CAS Number:
Molecular Weight:
170.48
EC Number:
MDL number:
UNSPSC Code:
12352300

vapor density

>1 (vs air)

grade

technical

Assay

≥98% (complexometric)

mp

100 °C (dec.) (lit.)

anion traces

sulfate (SO42-): ≤1000 mg/kg

cation traces

Fe: ≤500 mg/kg

SMILES string

Cl[Cu]Cl.[H]O[H].[H]O[H]

InChI

1S/2ClH.Cu.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2

InChI key

MPTQRFCYZCXJFQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service