Skip to Content
Merck
All Photos(1)

Documents

12584

Supelco

Dioctyl phenylphosphonate

Selectophore

Synonym(s):

DOPP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5P(O)[O(CH2)7CH3]2
CAS Number:
Molecular Weight:
382.52
Beilstein:
2948496
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

liquid

refractive index

n20/D 1.478 (lit.)

bp

207 °C/4 mmHg (lit.)

density

0.967 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCOP(=O)(OCCCCCCCC)c1ccccc1

InChI

1S/C22H39O3P/c1-3-5-7-9-11-16-20-24-26(23,22-18-14-13-15-19-22)25-21-17-12-10-8-6-4-2/h13-15,18-19H,3-12,16-17,20-21H2,1-2H3

InChI key

HAKMAMKAFTZXOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Dioctyl phenylphosphonate (DOPP) was used as plasticizer, in an experiment conducted for designing iron ion sensor based on functionalised ZnO (Zinc Oxide) nanorods. It was also used in Raman and UV-vis spectroscopic study, of polyaniline (PANI) membranes containing different amounts of a bulky lipophilic cationic additive, tridodecylmethylammonium chloride (TDMACl).

Other Notes

Plasticizer used in ion-selective electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.H. Griffiths et al.
Analyst, 97, 420-420 (1972)
Iron ion sensor based on functionalized ZnO nanorods.
Kimkeang K
Electroanalysis, 24 (3), 521-528 (2012)
Raman and UV-vis spectroscopic study of polyaniline membranes containing a bulky cationic additive.
Lindfors T
Journal of Electroanalytical Chemistry, 518 (2), 131-138 (2002)
K Oohira et al.
Biophysical chemistry, 61(1), 29-35 (1996-08-30)
Electric characteristics of two kinds of membranes in response to NaCl and quinine were theoretically studied; one membrane composed of polyvinyl chloride (PVC) and dioctylphenylphosphonate (DOPP), and the other a lipid/PVC/DOPP membrane containing PVC, DOPP and a negatively charged lipid.
Samah Hamze et al.
Nanomaterials (Basel, Switzerland), 10(7) (2020-07-02)
High-quality graphene is an especially promising carbon nanomaterial for developing nanofluids for enhancing heat transfer in fluid circulation systems. We report a complete study on few layer graphene (FLG) based nanofluids, including FLG synthesis, FLG-based nanofluid preparation, and their thermal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service