Skip to Content
Merck
All Photos(2)

Key Documents

11074

Supelco

Disperse Red 1

analytical standard

Synonym(s):

N-Ethyl-N-(2-hydroxyethyl)-4-(4-nitrophenylazo)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N4O3
CAS Number:
Molecular Weight:
314.34
Colour Index Number:
11110
Beilstein:
5353614
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

160-162 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCN(CCO)c1ccc(cc1)\N=N\c2ccc(cc2)[N+]([O-])=O

InChI

1S/C16H18N4O3/c1-2-19(11-12-21)15-7-3-13(4-8-15)17-18-14-5-9-16(10-6-14)20(22)23/h3-10,21H,2,11-12H2,1H3/b18-17+

InChI key

FOQABOMYTOFLPZ-ISLYRVAYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disperse Red 1 is an azo dye. Its mutagenic activity depends on its chemical structure.

Application

It was used as a dye in lymphocyte assay performed to evalute the formation of micronucleus in human lymphocytes.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

This benchmark dipolar chromophore was incorporated into aromatic polymers to obtain model electrooptic polymer films.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francine Inforçato Vacchi et al.
The Science of the total environment, 442, 302-309 (2012-11-28)
Azo dyes, the most widely used family of synthetic dyes, are often employed as colorants in areas such as textiles, plastics, foods/drugs/cosmetics, and electronics. Following their use in industrial applications, azo dyes have been found in effluents and various receiving
Mirosława Poprawa-Smoluch et al.
The journal of physical chemistry. A, 110(43), 11926-11937 (2006-10-27)
The photoisomerization of the push-pull substituted azo dye Disperse Red 1 is studied using femtosecond time-resolved absorption spectroscopy and other spectroscopic and computational techniques. In comparison with azobenzene, the pipi* state is more stabilized by the effects of push-pull substitution
An outbreak of occupational textile dye dermatitis from Disperse Blue 106.
F Mota et al.
Contact dermatitis, 43(4), 235-237 (2000-09-30)
Farah Maria Drumond Chequer et al.
Mutation research, 676(1-2), 83-86 (2009-05-16)
The use of azo dyes by different industries can cause direct and/or indirect effects on human and environmental health due to the discharge of industrial effluents that contain these toxic compounds. Several studies have demonstrated the genotoxic effects of various
Yaoquan Tu et al.
The journal of physical chemistry. B, 111(14), 3591-3598 (2007-03-29)
We demonstrate a complete procedure for simulations of electric field poled polymeric nonlinear optical systems with the purpose to evaluate the macroscopic electro-optic coefficients. The simulations cover the electric field poling effects on the chromophore order at the liquid state

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service