Skip to Content
Merck
All Photos(1)

Documents

09937

Sigma-Aldrich

Leit-Silver

suitable for electron microscopy

Synonym(s):

Conduct-Silver

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41111761
NACRES:
NA.23

technique(s)

electron microscopy: suitable

Quality Level

particle size

7-8 μm

storage temp.

2-8°C

General description

slow drying

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service