Skip to Content
Merck
All Photos(2)

Key Documents

09678

Millipore

Ammonium bismuth citrate

base ingredient for culture media

Synonym(s):

Citric acid ammonium bismuth salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€113.00
500 G
€379.00

€113.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€113.00
500 G
€379.00

About This Item

CAS Number:
EC Number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

€113.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

sterility

non-sterile

Assay

≥99.0% (KT)

form

powder

application(s)

microbiology

SMILES string

OC(CC(C(O)=O)(O)CC(O)=O)=O.C.C

InChI

1S/2C6H8O7.Bi.3H3N/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;3*1H3/q;;+3;;;/p-3

InChI key

VZZHAYFWMLLWGG-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium bismuth citrate is used in microbiological culture media as a selective agent as it inhibits certain gram-negative and gram-positive bacteria. The bismuth ions are reduced to metallic bismuth, which impart metallic sheen around the colonies.

Application

Ammonium bismuth citrate is used in microbiological culture media as a selective agent. It is for example used in Bismuth sulfite agar for the isolation of Salmonella typhi and other salmonellae from food and other materials, sewage and water. It is as well used in the Nickerson Agar (BIGGY), which is used for the isolation and identification of Candida species

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reduction of inorganic substances by yeasts I. Extracellular reduction of sulfite by species of Candida
Nickerson, Walter J
The Journal of Infectious Diseases, 43-56 (1953)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service