Skip to Content
Merck
All Photos(1)

Key Documents

09207

Supelco

cis-1,3-Pentadiene

analytical standard

Synonym(s):

cis-1-Methyl-1,3-butadiene, cis-Piperylene

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€295.00

€295.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€295.00

About This Item

Linear Formula:
CH3CH=CHCH=CH2
CAS Number:
Molecular Weight:
68.12
Beilstein:
1523658
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€295.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor pressure

6.58 psi ( 20 °C)

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.436 (lit.)
n20/D 1.437 (lit.)

bp

44 °C (lit.)

mp

−141 °C (lit.)

density

0.694 g/mL at 20 °C
0.691 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C\C=C/C=C

InChI

1S/C5H8/c1-3-5-4-2/h3-5H,1H2,2H3/b5-4-

InChI key

PMJHHCWVYXUKFD-PLNGDYQASA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service