Skip to Content
Merck
All Photos(1)

Key Documents

07966

Sigma-Aldrich

Methoxypolyethylene glycol amine

20,000

Synonym(s):

O-(2-Aminoethyl)-O′-methylpolyethylene glycol, Aminopolyethylene glycol monomethyl ether, Methoxypolyoxyethylene amine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€78.50

€78.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€78.50

About This Item

Linear Formula:
H2NCH2CH2(OCH2CH2)nOCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.22

€78.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

InChI

1S/C3H9NO/c1-5-3-2-4/h2-4H2,1H3

InChI key

ASUDFOJKTJLAIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vimala N Bharadwaj et al.
Nanomedicine : nanotechnology, biology, and medicine, 14(7), 2155-2166 (2018-06-23)
Clinically, traumatic brain injury (TBI) results in complex heterogeneous pathology that cannot be recapitulated in single pre-clinical animal model. Therefore, we focused on evaluating utility of nanoparticle (NP)-based therapeutics following three diffuse-TBI models: mildclosed-head injury (mCHI), repetitive-mCHI and midline-fluid percussion

Articles

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service