Skip to Content
Merck
All Photos(1)

Key Documents

07741

Supelco

Everolimus

analytical standard

Synonym(s):

CERTICAN®, 42-O-(2-Hydroxyethyl)rapamycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H83NO14
CAS Number:
Molecular Weight:
958.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

storage temp.

−20°C

SMILES string

O=C(C([C@@]1(O)[C@H](C)CC[C@](C[C@H](OC)/C(C)=C/C=C/C=C/[C@@H](C)C[C@@H](C)C2=O)([H])O1)=O)N3[C@H](C(O[C@@]([C@@H](C[C@@]4([H])C[C@@H](OC)[C@H](OCCO)CC4)C)([H])CC([C@H](C)/C=C(C)/[C@@H](O)[C@H]2OC)=O)=O)CCCC3

InChI

1S/C53H83NO14/c1-32-16-12-11-13-17-33(2)44(63-8)30-40-21-19-38(7)53(62,68-40)50(59)51(60)54-23-15-14-18-41(54)52(61)67-45(35(4)28-39-20-22-43(66-25-24-55)46(29-39)64-9)31-42(56)34(3)27-37(6)48(58)49(65-10)47(57)36(5)26-32/h11-13,16-17,27,32,34-36,38-41,43-46,48-49,55,58,62H,14-15,18-26,28-31H2,1-10H3/b13-11+,16-12+,33-17+,37-27+/t32-,34-,35-,36-,38-,39+,40+,41+,43-,44+,45+,46-,48-,49+,53-/m1/s1

InChI key

HKVAMNSJSFKALM-GKUWKFKPSA-N

Gene Information

human ... FKBP1A(2280)

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

Exact concentration and expiry date can be found on the certificate, included in each package.

Legal Information

CERTICAN is a registered trademark of Novartis AG

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Kidney,Testes

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsushi Ohtsu et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(31), 3935-3943 (2013-09-18)
The oral mammalian target of rapamycin inhibitor everolimus demonstrated promising efficacy in a phase II study of pretreated advanced gastric cancer. This international, double-blind, phase III study compared everolimus efficacy and safety with that of best supportive care (BSC) in
Joëlle Elias et al.
Heart (British Cardiac Society), 104(17), 1432-1438 (2018-02-22)
During primary percutaneous coronary intervention (PCI), a concurrent chronic total occlusion (CTO) is found in 10% of patients with ST-elevation myocardial infarction (STEMI). Long-term benefits of CTO-PCI have been suggested; however, randomised data are lacking. Our aim was to determine
Robert J Motzer et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 32(25), 2765-2772 (2014-07-23)
A multicenter, randomized phase II trial, RECORD-3, was conducted to compare first-line everolimus followed by sunitinib at progression with the standard sequence of first-line sunitinib followed by everolimus in patients with metastatic renal cell carcinoma. RECORD-3 used a crossover treatment
Inge Verbrugge et al.
Radiation research, 182(2), 219-229 (2014-06-25)
Radiotherapy is a successful treatment modality for localized cancer. Our group has been exploring radiotherapy in combination with immunotherapy (radioimmunotherapy) to enhance systemic antitumor responses. Previously, we have shown that when local radiotherapy was combined with monoclonal antibodies (mAbs) (that
Yvonne Yeung et al.
Molecular oncology, 11(9), 1130-1142 (2017-05-26)
Advanced biliary tract cancer (BTC) has a poor prognosis and limited treatment options. The PI3K/Akt/mTOR signalling pathway is hyperactivated in a subset of BTCs, and clinical activity of the mTOR inhibitor everolimus has been observed in some patients with BTC.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service