Skip to Content
Merck
All Photos(1)

Key Documents

07512

Supelco

α-Hederin

analytical standard

Synonym(s):

(3β,4α)-3-[[2-O-(6-Deoxy-α-L-mannopyranosyl)-α-L-arabinopyranosyl]oxy]-23-hydroxyolean-12-en-28-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H66O12
CAS Number:
Molecular Weight:
750.96
EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤10.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@@H](O)CO[C@H]2O[C@H]3CC[C@@]4(C)[C@@H](CC[C@]5(C)[C@@H]4CC=C6[C@@H]7CC(C)(C)CC[C@@]7(CC[C@@]56C)C(O)=O)[C@]3(C)CO)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C41H66O12/c1-21-28(44)30(46)31(47)33(51-21)53-32-29(45)24(43)19-50-34(32)52-27-11-12-37(4)25(38(27,5)20-42)10-13-40(7)26(37)9-8-22-23-18-36(2,3)14-16-41(23,35(48)49)17-15-39(22,40)6/h8,21,23-34,42-47H,9-20H2,1-7H3,(H,48,49)/t21-,23-,24-,25+,26+,27-,28-,29-,30+,31+,32+,33-,34-,37-,38-,39+,40+,41-/m0/s1

InChI key

KEOITPILCOILGM-LLJOFIFVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Hederin is a water-soluble pentacyclic triterpene compound present in the seeds of Nigella sativa.

Application

α-Hederin may be used as an internal standard for the determination of glycyrrhizin and its major metabolite glycyrrhetic acid in human plasma by liquid chromatography/tandem mass spectrometry (LC−MS/MS). It may be used as an external standard for quinovic acid glycosides assay by high performance liquid chromatography-photodiode array detection (HPLC-PDA) as well as ultra-performance liquid chromatography-quadrupole time-of-flight mass spectrometry (UPLC/Q-TOF-MS). It may be used as a reference standard for the identification of α-hederin in the leaves of common ivy (Hedera helix) by liquid chromatography-electrospray ionization tandem mass spectrometry (LC-EI/MS-MS).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Preclinical and clinical effects of Nigella sativa and its constituent, thymoquinone: A review.
Gholamnezhad Z, et al.
Journal of Ethnopharmacology, 190, 372-386 (2016)
Simultaneous determination of glycyrrhizin, a marker component in radix Glycyrrhizae, and its major metabolite glycyrrhetic acid in human plasma by LC-MS/MS.
Lin Z, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 814(2), 201-207 (2005)
HPLC-PDA method for quinovic acid glycosides assay in Cat's claw (Uncaria tomentosa) associated with UPLC/Q-TOF-MS analysis.
Pavei C, et al.
Journal of Pharmaceutical and Biomedical Analysis, 62(2), 250-257 (2012)
Yang Liu et al.
Molecules (Basel, Switzerland), 15(11), 7871-7883 (2010-11-10)
Thirteen novel triterpenoid saponins, designed as amide derivatives of the natural cytotoxic saponin β-hederin, were synthesized by a stepwise glycosylation strategy. The in vitro cytotoxic activity of these compounds was evaluated against five different tumor cell lines. Most of the
Anne Wolf et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(2-3), 214-218 (2010-07-20)
Preparations of ivy leaves dry extract with secretolytic and bronchiolytic efficacy are widely used for the treatment of acute and chronic obstructive airway diseases. The mechanism by which ivy preparations improve lung functions is not fully understood. Here, we tested

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service