Skip to Content
Merck
All Photos(1)

Documents

06571

Supelco

Amine ionophore I

Selectophore, function tested

Synonym(s):

Calix[6]arene-hexaacetic acid hexaethylester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H72O18
CAS Number:
Molecular Weight:
1153.27
Beilstein:
5231462
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

form

powder

quality

function tested

SMILES string

CCOC(=O)COc1c2Cc3cccc(Cc4cccc(Cc5cccc(Cc6cccc(Cc7cccc(Cc1ccc2)c7OCC(=O)OCC)c6OCC(=O)OCC)c5OCC(=O)OCC)c4OCC(=O)OCC)c3OCC(=O)OCC

InChI

1S/C66H72O18/c1-7-73-55(67)37-79-61-43-19-13-20-44(61)32-46-22-15-24-48(63(46)81-39-57(69)75-9-3)34-50-26-17-28-52(65(50)83-41-59(71)77-11-5)36-54-30-18-29-53(66(54)84-42-60(72)78-12-6)35-51-27-16-25-49(64(51)82-40-58(70)76-10-4)33-47-23-14-21-45(31-43)62(47)80-38-56(68)74-8-2/h13-30H,7-12,31-42H2,1-6H3

InChI key

LSVIETSWDFUREK-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ionophore used in polymeric membrane electrodes selective for primary amines

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service