Skip to Content
Merck
All Photos(1)

Key Documents

06457

Supelco

Amberlite IRA910 Ion Exchange Resin

chloride form, 16-50 mesh, strongly basic

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€144.00

€144.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
€144.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

€144.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Amberlite IRA910 chloride form, chloride form, strongly basic

form

beads

parameter

40 °C OH- form max. temp.
77 °C Cl- form max. temp.

moisture

54-61%

technique(s)

LPLC: suitable

loss

~55% loss on drying

matrix

styrene-divinylbenzene (macroreticular)

matrix active group

dimethylethanolamine functional group

particle size

530-800 μm

operating pH

0-14

capacity

1.0 meq/mL by wetted bed volume

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

Application

Strongly basic (type II) anion exchange resin for water conditioning and deionization, removal of sulfuric acid and color, etc. The type II base provides improved regeneration efficiency over type I base resin.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service