Skip to Content
Merck
All Photos(1)

Key Documents

06369

Sigma-Aldrich

Lipid binding constant determination kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

contains

0.0005% NaN3 as preservative

concentration

30-35 mM lipid (Transil®-L pellet)

surface area

~10 m2/g

particle size

~30 μm

pore size

~4000 Å pore diameter

storage temp.

2-8°C

Application

Lipid binding constant determination kit is used for analysis of lipid binding.

Components

TRANSIL®-Ln (neutral biomembrane);
1 mL TRANSIL®-Lc (biomembrane with negative charge clusters); 1 ml
each in 5 mL buffer (20 mM HEPES pH 7.0; 0.03 M NaCl)
TRANSIL®-L consists of a silicate core; encased in a mobile lipid bilayer (phospholipids).

Legal Information

Transil is a registered trademark of Nimbus GmbH

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Loidl-Stahlhofen et al.
Pharmaceutical research, 18(12), 1782-1788 (2002-01-12)
Lipid-water partitioning of 187 pharmaceuticals has been assessed with solid-supported lipid membranes (TRANSIL) in microwell plates and with multilamellar liposomes for a data comparison. The high-throughput potential of the new approach was evaluated. Drugs were incubated at pH 7.4 with
A Loidl-Stahlhofen et al.
Journal of pharmaceutical sciences, 90(5), 599-606 (2001-04-05)
Quantification of membrane affinity is an important early screening step in modern drug design. However, current approaches using different lipid membrane models usually are time-consuming or show severe experimental drawbacks. In this paper we describe the use of solid-supported lipid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service