Skip to Content
Merck
All Photos(1)

Documents

04642

Supelco

3-Acetylpyridine

analytical standard

Synonym(s):

Methyl 3-pyridyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
Beilstein:
107751
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.534 (lit.)
n20/D 1.535

bp

220 °C (lit.)

mp

11-13 °C (lit.)

density

1.102 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)c1cccnc1

InChI

1S/C7H7NO/c1-6(9)7-3-2-4-8-5-7/h2-5H,1H3

InChI key

WEGYGNROSJDEIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may discolor to deep yellow on storage

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huawen Lin et al.
PLoS genetics, 6(9), e1001105-e1001105 (2010-09-15)
The essential coenzyme nicotinamide adenine dinucleotide (NAD+) plays important roles in metabolic reactions and cell regulation in all organisms. Bacteria, fungi, plants, and animals use different pathways to synthesize NAD+. Our molecular and genetic data demonstrate that in the unicellular
Iran Goudarzi et al.
European journal of pharmacology, 642(1-3), 56-65 (2010-07-20)
Electrophysiological dysfunction of Purkinje cells causes cerebellar ataxia. Recent studies indicated that 4-aminopyridine (4-AP) can prevent the attacks in patients with episodic ataxia type 2. However, the cellular mechanism(s) by which 4-AP might be beneficial for the improvement of motor
M Kaffashian et al.
Physiological research, 60(2), 355-365 (2010-12-01)
Alterations in the intrinsic properties of Purkinje cells (PCs) may contribute to the abnormal motor performance observed in ataxic rats. To investigate whether such changes in the intrinsic neuronal excitability could be attributed to the role of Ca(2+)-activated K(+) channels
Ai-Li Cai et al.
The European journal of neuroscience, 24(8), 2169-2176 (2006-10-18)
Zinc neurotoxicity has been demonstrated in ischemic, seizure, hypoglycemic, and trauma-induced neuronal death where Zn(2+) is thought to be synaptically released and taken up in neighbouring neurons, reaching toxic concentrations. We previously demonstrated that toxicity of extracellular Zn(2+) depended on
Amanda Sierra et al.
Endocrine, 21(1), 43-51 (2003-06-05)
The expression of aromatase, the enzyme that transforms testosterone into estradiol, was analyzed by reverse transcriptase polymerase chain reaction in the inferior olive of adult male rats. The expression of this messenger in the inferior olive suggests that this brain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service