Skip to Content
Merck
All Photos(1)

Documents

04304

Supelco

9-(Hydroxymethyl)-10-carbamoylacridan

analytical standard

Synonym(s):

9-HMCA, 9-OH-CBZ, 9-(Hydroxymethyl)-10(9H)-acridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N2O2
CAS Number:
Molecular Weight:
254.28
Beilstein:
6214413
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

NC(N1C2=CC=CC=C2C(CO)C3=C1C=CC=C3)=O

InChI

1S/C15H14N2O2/c16-15(19)17-13-7-3-1-5-10(13)12(9-18)11-6-2-4-8-14(11)17/h1-5,7-8,18H,6,9H2,(H2,16,19)

InChI key

UMRKOEWAECPINL-UHFFFAOYSA-N

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arnold Bahlmann et al.
Water research, 57, 104-114 (2014-04-08)
The occurrence of carbamazepine (CBZ) and its metabolites in German and Portuguese wastewater was investigated. A total of 46 samples from influent and effluent wastewater were analyzed by liquid-chromatography (LC) tandem mass spectrometry. The five metabolites 10,11-dihydro-10,11-dihydroxy-CBZ (DiOH-CBZ), 10,11-dihydro-10-hydroxy-CBZ (10-OH-CBZ)
The metabolism of carbamazepine in humans: steric course of the enzymic hydrolysis of the 10, 11-epoxide
Bellucci G, et al.
Journal of Medicinal Chemistry, 30(5), 768-773 (1987)
9-Hydroxymethyl-10-carbamoylacridan in human serum is one of the major metabolites of carbamazepine
Seiji ETO, et al.
Biological & Pharmaceutical Bulletin, 18(6), 926-928 (1995)
S Eto et al.
Biological & pharmaceutical bulletin, 18(6), 926-928 (1995-06-01)
In sera of 29 patients under the chronic administration of carbamazepine (CBZ, dose: 10.9 +/- 4.3 mg/kg/day), a ring-contracted metabolite of CBZ was detected at a relatively higher concentration of 0.57 +/- 0.30 mu g/ml (mean +/- S.D.) by high-performance
Carbamazepine: detection of another metabolite in serum, 9 hydroxymethyl-10-carbamoyl acridan
Wad N, et al.
Therapeutic Drug Monitoring, 19(3), 314-317 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service