Skip to Content
Merck
All Photos(1)

Key Documents

03650

Sigma-Aldrich

Ethylenediaminetetraacetic acid iron(III) sodium salt hydrate

12-14% Fe basis

Synonym(s):

EDTA iron(III) sodium salt, Iron(III)-sodium ethylenediaminetetraacetate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€60.40
1 KG
€209.00

€60.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€60.40
1 KG
€209.00

About This Item

Linear Formula:
(OOCCH2)2NCH2CH2N(CH2COO)2FeNa · xH2O
CAS Number:
Molecular Weight:
367.05
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes
form:
powder or crystals
solubility:
hot water: 0.5 g/10 mL, clear to slightly hazy (Yellow to Very Dark Yellow to Very Dark Brown)

€60.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

12-14% (Fe basis)

form

powder or crystals

composition

Fe, 12-14%

reaction suitability

reagent type: chelator

impurities

≤15.0% Loss on drying

solubility

hot water: 0.5 g/10 mL, clear to slightly hazy (Yellow to Very Dark Yellow to Very Dark Brown)

SMILES string

[Na+].[Fe+3].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.Fe.Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;+3;+1/p-4

InChI key

MKWYFZFMAMBPQK-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Ethylenediaminetetraacetic acid iron(III) sodium salt hydrate is used in large-volume stacking with electroosmotic flow pump (LVSEP)[1] and in the remediation of contaminated groundwater.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidative and reductive pathways in iron-ethylenediaminetetraacetic acid?activated persulfate systems.
Ahmad M, et al.
Journal of Environmental Engineering (New York, N.Y.), 138(4), 411-418 (2011)
Large?volume sample stacking for analysis of ethylenediaminetetraacetic acid by capillary electrophoresis.
Zhu Z, et al.
Electrophoresis, 23(17), 2880-2887 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service