Skip to Content
Merck
All Photos(1)

Key Documents

03560

Sigma-Aldrich

Ethylenediamine

purum, ≥99.0% (GC)

Synonym(s):

1,2-Diaminoethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NH2
CAS Number:
Molecular Weight:
60.10
Beilstein:
605263
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

vapor density

2.07 (vs air)

vapor pressure

10 mmHg ( 20 °C)

grade

purum

Assay

≥99.0% (GC)

autoignition temp.

716 °F

expl. lim.

16 %

impurities

≤1% water

refractive index

n20/D 1.4565 (lit.)
n20/D 1.457

bp

118 °C (lit.)

mp

8.5 °C (lit.)

density

0.899 g/mL at 25 °C (lit.)

SMILES string

NCCN

InChI

1S/C2H8N2/c3-1-2-4/h1-4H2

InChI key

PIICEJLVQHRZGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethylenediamine solution was used in the synthesis of ultra-small CdSe quantum dots.[1] It may be used as solvent in the preparation of ZnS and ZnSe precursors (ZnS·0.5en and ZnSe·0.5en, en = ethylenediamine), via solvothermal process.[2] It may be employed for the synthesis of inulin based polycation (Inu-EDA).[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mild synthesis of ultra-small CdSe quantum dots in ethylenediamine solution.
Yoon J-H, et al.
Materials Letters, 59(11), 1430-1433 (2005)
Absorption of carbon dioxide into aqueous solutions of ethylenediamine: Effect of interfacial turbulence.
Sada E, et al.
Chemical Engineering Journal, 13(3), 213-217 (1977)
Zhao-Xiang Deng et al.
Inorganic chemistry, 41(4), 869-873 (2002-02-19)
ZnS and ZnSe precursors (ZnS x 0.5en and ZnSe x 0.5en, en = ethylenediamine) were prepared via a solvothermal process using ethylenediamine as solvent. Phase-pure hexagonal wurtzite ZnS and ZnSe products were obtained by annealing the precursors in argon stream
Mariano Licciardi et al.
Pharmaceutical research, 32(11), 3674-3687 (2015-06-19)
An inulin based polycation (Inu-EDA) has been synthesized by the grafting of ethylenediamine molecules onto inulin backbone. The obtained inulin copolymer has been though to coat SPIONs (IC-SPIONs) and obtain stable magnetoplexes by complexation of IC-SPIONs with a model duplexed
Ana Carolina M S Dias et al.
Biosensors & bioelectronics, 44, 216-221 (2013-02-23)
An immunosensor for the non-structural protein 1 (NS1) of the dengue virus based on carbon nanotube-screen printed electrodes (CNT-SPE) was successfully developed. A homogeneous mixture containing carboxylated carbon nanotubes was dispersed in carbon ink to prepare a screen printed working

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service