Skip to Content
Merck
All Photos(1)

Documents

02558

Supelco

Rhodamine B solution

0.2% in isopropanol, for TLC derivatization

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31ClN2O3
CAS Number:
Molecular Weight:
479.01
Beilstein:
4119648
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for TLC derivatization

Quality Level

concentration

0.2% in isopropanol

technique(s)

thin layer chromatography (TLC): suitable

density

0.79 g/mL at 20 °C

SMILES string

[Cl-].CCN(CC)c1ccc2c(OC3=CC(\C=CC3=C2c4ccccc4C(O)=O)=[N+](\CC)CC)c1

InChI

1S/C28H30N2O3.ClH/c1-5-29(6-2)19-13-15-23-25(17-19)33-26-18-20(30(7-3)8-4)14-16-24(26)27(23)21-11-9-10-12-22(21)28(31)32;/h9-18H,5-8H2,1-4H3;1H

InChI key

PYWVYCXTNDRMGF-UHFFFAOYSA-N

Application

ready-to-use spray and dip reagent for chromatography

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda Johansson et al.
Ultrasonics, 53(5), 1020-1032 (2013-03-19)
An acoustic trap with miniaturized integrated transducers (MITs) for applications in non-contact trapping of cells or particles in a microfluidic channel was characterized by measuring the temperature increase and trapping strength. The fluid temperature was measured by the fluorescent response
Van Duong Ta et al.
Scientific reports, 3, 1362-1362 (2013-03-02)
Optical microcavities are important for both fundamental studies of light-matter interaction and applications such as microlasers, optical switches and filters etc... Tunable microresonators, in which resonant modes can be manipulated, are especially fascinating. Here we demonstrate a unique approach to
Shengnan Su et al.
Journal of hazardous materials, 244-245, 736-742 (2012-12-01)
The removal of Rhodamine B (RhB) by Co(x)Fe(3-x)O(4) magnetic nanoparticles activated Oxone has been performed in this study. A series of Co(x)Fe(3-x)O(4) nanoparticles was synthesized using a hydrothermal method. The synthetic Co(x)Fe(3-x)O(4) nanoparticles were characterized using X-ray diffraction (XRD) and
Qi Wang et al.
Journal of hazardous materials, 246-247, 135-144 (2013-01-10)
Amorphous TiO(2) (Am-TiO(2)) was prepared at room temperature by hydrolysis of Ti(OBu)(4) in water without addition of strong acids or organic solvents. Results from XRD and TEM revealed that the as-prepared Am-TiO(2) was composed of amorphous structure. For the simultaneous
Daniel J Coles et al.
Chemical communications (Cambridge, England), 49(37), 3836-3838 (2013-02-19)
Hyperbranched polymers conjugated to a peptide-aptamer were prepared using a combination of RAFT polymerisation and click chemistry for targeting tumour cells in vivo. The polymers showed enhanced cell-uptake in vitro (compared to unconjugated polymer) while excellent specificity for solid tumours

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service